Temperature Limit Control, Safety Device, Ace Glass
Fournisseur: Ace Glass
Certificats
A propos de cet article
Single outlet, secondary temperature control safety device for applications where temperature overshoot will cause dangerous conditions
- Upon being tripped, an audible alarm will sound, and a manual reset will be required
Used as a power source for your in-house temperature controller, the limit control will cut power to its outlet when your upper temperature setpoint is reached.
Ordering information: Includes integral rack mounting bracket. One-year limited warranty.
Spécifications
- Catalog No:
- 75846-436
- 75846-438
- 75846-440
- 75846-442
- 75846-432
- 75846-446
- 75846-750
- 75846-444
- Accuracy:
- 0.1% of span, ± 1.0°C
- 0.1% of span, ± 1.0°C
- 0.1% of span, ± 1.0°C
- 0.1% of span, ± 1.0°C
- 0.1% of span, ± 1.0°C
- 0.1% of span, ± 1.0°C
- 0.1% of span, ± 1.0°C
- 0.1% of span, ± 1.0°C
- Electrical:
- 120V, 15 Amp, 60Hz input; 1800W output
- 120V, 15 Amp, 60Hz input; 1800W output
- 120V, 15 Amp, 60Hz input; 1800W output
- 120V, 15 Amp, 60Hz input; 1800W output
- 120V, 15 Amp, 60Hz input; 1800W output
- 120V, 15 Amp, 60Hz input; 1800W output
- 120V, 15 Amp, 60Hz input; 1800W output
- 120V, 15 Amp, 60Hz input; 1800W output