Accelerator Free, Class 100 Nitrile Cleanroom Glove, TechNiGlove
Supplier: Techniglove
Certificates
About this item
Rival CR's new soft modulus nitrile delivers the fit and feel of latex for the ultimate stretch and comfort.
- 100% Nitrile
- Powder-free
- Accelerator-free
- Soft modulus
Rival CR 12" ambidextrous Class 100 Nitrile gloves are accelerator-free for those with skin sensitivities. TechNiGlove products contain no natural rubber or silicone, and meet the most stringent requirements for extractables and particles.
Suitable for cleanroom, pharmaceutical, biotech, and medical device manufacturing.
Packaging: Packaged 100 gloves per double poly-sealed bag, 10 bags per case.
Specifications
- Catalog No:
- 75845-756
- 75845-758
- 75845-748
- 75845-750
- 75845-754
- 75845-752
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Powdered/Powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Finish:
- Microtextured Fingertips
- Microtextured Fingertips
- Microtextured Fingertips
- Microtextured Fingertips
- Microtextured Fingertips
- Microtextured Fingertips