Water-Filled Boom Diverter, Justrite®
Fournisseur: Justrite
Certificats
A propos de cet article
Temporarily divert and control fluid for easy cleanup or recovery.
- Helps comply with EPA for stormwater regulations
- Redirects liquids away from work areas and storm drains
Use the water filled booms in storm water management, portable washdown/pressure washing diversion, spill diversion, and equipment and personnel decontamination. Flexible PVC film construction conforms to uneven surfaces such as asphalt or concrete to form a diversionary barrier away from storm drains or as a holding area for wet vacuum operations. Features easy-open push plug for filling with standard water hose.
Customers Who Bought This Also Bought