Ward's® Biology Flashcards
# de catalogue 470022-908
Fournisseur: MERCURY PRINT PRODUCTIONS SE
Spécifications
- Description:Evolution Flash Cards, 75 Cards
- Cat. no.:470022-908
- Length:12.7 cm
- Width:7.6 cm
- Supplier No.:323392
Spécifications
A propos de cet article
Perfect Review Tool For Exams
Each set of cards contains a mixture of terms, illustrations, definitions, and simple problems.
Customers Who Bought This Also Bought