Sanitary Gaskets, PTFE/Elastomer Core, Chemglass
Supplier: Chemglass
Certificates
About this item
Gasket grafts a PTFE envelope to an EPDM elastomer core.
- Gasket will not creep or cold flow overtime so it maintians the integrity of its seal
- Temperature range −73 to +177 °C
Customers Who Bought This Also Bought