Gas-Tight Micro Syringes, Chemglass
Fournisseur: Chemglass
Certificats
A propos de cet article
These syringes are designed for use in both liquid and gas chromatography.
- Interchangeable within each size range
- Available in capacities from 5µL to 50mL
Due to the all-PTFE tip, they can also handle highly corrosive or Ultra-pure liquids.
Delivery: Fixed and removable needle syringes are supplied with needles that are 2" long with point style "A" (22° bevel).
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.