NMR Tube Cleaner, Wong, Chemglass
Fournisseur: Chemglass
Certificats
A propos de cet article
It permits the cleaning of four NMR tubes at the same time.
- With solvent in the reservoir, rapid suction wets the inside of the NMR tube dislodging the contamination
- An acetone wash followed by an air stream dries the tube in seconds
- Rate of cleaning is approximately four tubes per minute
The NMR tube is inserted into the cleaner with a poly-pro cap placed on the closed end to provide support and a vacuum seal. Lower joint is a 24/40 inner, and the hose connection has an Ext.Ø of 8.5 mm at the middle serration. Complete unit comes with an NMR tube washer and 250 ml heavy wall filter flask.
Customers Who Bought This Also Bought