SP Wilmad-LabGlass Capillary Melting Point Tubes, SP Industries
Fournisseur: SP Industries
Certificats
A propos de cet article
Fabricated from low extractable borosilicate glass, these melting point capillary tubes will not react with organic compounds.
Capillary tubes are closed at one end and packed in vials containing approximately 100 tubes.
Customers Who Bought This Also Bought