Airgas® Single Stage Brass Acetylene Cylinder Regulator
Fournisseur: Airgas
Certificats
A propos de cet article
Single stage gas regulators are engineered to provide precision performance.
- Filtered seat for added gas stream purity and extended service life
- Reverse flow check valve prevents the reverse flow of gas into the regulator
- Large, reinforced diaphragm provides excelent performance characteristics
- Red warning delivery gauge indicates delivery pressure exceeding 15 psig
The regulators feature a 10-micron, sintered stainless steel filter and a PTFE-lined, nylon-reinforced neoprene diaphragm.
Spécifications
- Catalog No:
- 47752-000
- 47752-002
- Max. inlet pressure:
- 400 psig
- 400 psig
- Outlet pressure:
- 0 - 15 psig
- 0 - 15 psig
- Inlet/outlet:
- 1/4" FNPT/9/16-18 LH
- 1/4" FNPT/9/16-18 LH