FIREKING® Lateral File Cabinets, Essendant LLC MS
# de catalogue 470116-204
Fournisseur: Fireking International Inc
Spécifications
- Depth:56.2 cm (22 1/8")
- Width:79.2 cm (31 3/16")
- Height:134 cm (52 3/4")
- Cat. no.:470116-204
- Weight:360 kg (794 lbs.)
- Supplier No.:4-3122-C
Spécifications
A propos de cet article
Lateral File Cabinet is equipped to handle both letter and legal hanging files, no additional frames are required.
- High security key lock
- Water resistant
- Four drawers
Certifications: UL Class 350 1-hour fire and impact rating.
Customers Who Bought This Also Bought