WHEATON® Filtering Flask
# de catalogue 26316-558
Fournisseur: DWK Life Sciences
Small Business Enterprise
Spécifications
- Capacity:1000 ml
- Description:Filtering Flask for HPLC Solvents
- Cat. no.:26316-558
- Supplier No.:635527
Spécifications
A propos de cet article
Filtering flask is used for the filtration of organic solvents, corrosive liquids and the removal of particulates from HPLC solvents.
- Recommended for general filtration
- Units have a #2 hose connection for 1/4# (6mm) I.D. flexible tubing
- Ground glass connection eliminates phthalate contamination from silicone stopper
Certifications: Glass components are low extractable borosilicate glass that conforms to USP Type I and ASTM E 438 Type I, Class A requirement
Delivery: Includes a coarse porosity (40-60µm) fritted glass support base, graduated funnel, anodized aluminum clamp and 1 L filter flask
Customers Who Bought This Also Bought