Magnetizer Box
# de catalogue 470159-586
Fournisseur: Wards
Spécifications
- Description:Magnetizer Box
- Cat. no.:470159-586
- Supplier No.:MAGBAR-V1
Spécifications
A propos de cet article
No need to Buy New Magnets
- Bar Magnet
- 110/120V AC
- 60Hz
Revitalizes iron, nickel, cobalt, and alnico horseshoe and bar magnets in seconds. Simply place magnets in the 25x25 mm holes and press the button to make old magnets new again.
Customers Who Bought This Also Bought