Foamtips™ Foam Swabs, ITW Chemtronics®
# de catalogue 15145-508
Fournisseur: ITW Chemtronics
Spécifications
- Description:Foamtips™ 100 ppi open cell foam with PP handle
- Cat. no.:15145-508
- Overall Length:130 mm (5.12")
- Head Width:12.7 mm (0.50")
- Head Length:25.4 mm (1")
- Handle Width:4.8 mm (0.19")
- Supplier No.:CF2050
Spécifications
A propos de cet article
The Foamtips™ swabs are constructed from purest medical grade foam which features the best open cell structure, providing excellent particle entrapment characteristics. The foam is thermally bonded to the swab handle without using adhesives. These swabs are ideal for general cleaning applications.
- Precision cleaning swabs with polyurethane foam head
- Traps surface particles while cleaning
- High solvent capacity, holds solvent well
- Fiberless construction does not generate loose fibers or particles
- No adhesives or binders are used in the construction
- Economical for high quantity applications
It is widely used for cleaning excess adhesives after gluing, micro-mechanical devices, removes flux residues from printed circuit boards and used as adhesive or lubricants.
Customers Who Bought This Also Bought