Holder for Rugged Rotator, Glassware Variety, Glas-Col
Catalog # 57020-265
Supplier: Glas-Col
Small Business Enterprise
Specifications
- Capacity:Glassware up to dia. 2¼"
- Description:Clamping Head, Glassware Variety
- Cat. no.:57020-265
- Weight:5.5 lbs.
- Supplier No.:099ARD20
Specifications
About this item
Used with the Rugged Rotator for rotating and assortment of glassware.
- Mounts quickly on Rotator
- Clamps hold securely with no scratching
- Clamp position is adjustable
Equipped with six large 3-finger clamps which attach to the casting individually with lockable thumb screws. Clamp position adjusts to allow axial or end over end rotation.
Ordering information: Casting with 6 finger clamps
Delivery: 7 to 10 days after order.
Customers Who Bought This Also Bought