384-Well COC Compound Storage Microplate for Acoustic Liquid Handling, Greiner Bio-One
Catalog # 89800-680
Supplier: Greiner Bio-One
Specifications
- Color:Clear
- Description:384 Well COC Compound Storage Microplate
- Material:Cycloolefin
- Cat. no.:89800-680
- Working Volume:1 - 25 µl
- Supplier No.:793855
- Total Volume:28 µl
Specifications
About this item
For Compound Storage.
- Solid bottom
- Low dead volume
- Free of detectable DNase/RNase, human DNA and pyrogens
Microplates are deionised and packed in antistatic bags.
Frequently Bought Together