Econowipe Cleanroom Wipes, Liberty Industries
# de catalogue 21924-470
Fournisseur: Liberty Industries
Small Business Enterprise
Spécifications
- Dimensions:22.8 x 22.8 cm (9 x 9")
- Cat. no.:21924-470
- Packaging:300/Tub, 6 Tubs/Case
- Supplier No.:50-5120
Spécifications
A propos de cet article
These cleanroom wipes are perfect for light to medium duty wiping applications.
- Will not break down when wet
- Will absorb many times its own weight in liquid
Packaged in sturdy cleanroom dispenser plastic tubs that are ideal for clean storage and easy use. Monitored and tested to insure low levels of particulate matter, sodium and other heavy metals.
Customers Who Bought This Also Bought