Holder for Nutating Shakers, Erlenmeyer Flask, Glas-Col
Supplier: Glas-Col
Certificates
About this item
Specially designed for our floor model and mid range shakers
- Quick release front clamp for ease in filling of funnel
- Mount to shaker platform with no fasteners
These erlenmeyer flask holders have a rear support which remains stationary while front part holding the funnel can be released and rotated 360 degrees. This permits the funnel to be filled, inverted for shaking, and then reinverted for addition and/or extraction. All while the flask remains in the holder.
Ordering information: One holder holds one flask.
Delivery: 7-10 days after order.
Customers Who Bought This Also Bought