Holder for Bench Top Reciprocating Shaker, Erlenmeyer Flask, Glas-Col
# de catalogue 57020-289
Fournisseur: Glas-Col
Small Business Enterprise
Spécifications
- Capacity:125 ml, 250 ml or 500 ml
- Description:Flask Holder Rack for Erlenmeyer Flask
- Cat. no.:57020-289
- Supplier No.:099AEF521
Spécifications
A propos de cet article
This flask holder lets you shake one to four Erlenmeyer flasks at one time on our Bench Top Shaker.
- Easy adjustment accomodates one to four flasks
- Locking knob secures holder in optimum positions for loading and effective shaking
- Holder can be rotated 180 degrees while mounted on shaker
- Two flask holder can be shaken at once
Uncapped flasks can be shaken back and forth gently or you can close the flasks when a more vigorous back and forth motion is needed.
Ordering information: Accomodates three 500 mL flasks or four 125 mL to 250 L flasks.
Delivery: 7-10 days after order.
Customers Who Bought This Also Bought