CELLSTAR® 96-Well Cell Culture Microplate, Poly-D-Lysine, Greiner Bio-One
Catalog # 89093-212
Supplier: Greiner Bio-One
Product available on GSA Advantage®
Specifications
- Description:Microplate without Lid
- Cat. no.:89093-212
- Supplier No.:675946
Specifications
About this item
Half-area. Black color.
- µClear® bottom style
- PS material
Customers Who Bought This Also Bought