Circular Reaction Blocks, Chemglass
Fournisseur: Chemglass
Certificats
A propos de cet article
Round reaction blocks are designed to fit directly on 135 mm diameter, round top hot plate stirrers.
- Excellent heat transfer
- Holes spaced for optimum stirring of all vials
- Color-anodized for durability and easy identification
Aluminum construction offers excellent heat transfer without interfering with magnetic stirring. A thermowell is provided for insertion of a temperature sensor or contact thermometer.
Customers Who Bought This Also Bought