AIRLESS-WARE® Schlenk Vacuum Tubes, Kimble Chase
Fournisseur: DWK Life Sciences (KIMBLE)
Certificats
A propos de cet article
This Schlenk tube is suitable for vacuum gas manifolds with HI-VAC® valve to control gas flow to the reaction tube.
- Tubes are constructed of medium wall borosilicate glass
- Performs under both thermal and pressure-type reactions
Manufactured from 33 expansion, low extractable borosilicate glass.
The 25 and 50 ml tubes have 0 to 4 mm valves with 826601-0004 plugs.
The 100 and 350 ml tubes have 0 to 8 mm valves with 826601-0008 plugs.
Certifications: Conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Customers Who Bought This Also Bought