Roller Apparatus, Wheaton®, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
Certificats
A propos de cet article
Roller apparatus is used for the large scale cell culture.
- Robust touch screen interface that is easy to view from a distance
- Readily recognized icons for intuitive navigation
- Digitally controlled maintenance free motor accurate to 0.01 RPM
- Capable of remote control and monitoring through SCADA systems
- Multiple decking and capacity configurations available for process flexibility
- USB Downloadable System Event Log
Spécifications
- Catalog No:
- 89369-082
- 89369-080
- 89369-124
- 89368-998
- 89369-086
- 89369-084
- 89369-074
- 89369-118
- 89369-072
- 89369-116
- 89369-078
- 89369-122
- 89369-076
- 89369-120
- 89369-110
- 89369-070
- 89369-114
- 89369-112
- 10159-632
- 89369-104
- 89369-102
- 89369-108
- 89369-106
- 89369-096
- 89369-094
- 89369-012
- 89369-100
- 89369-010
- 89369-098
- 89369-092
- 89369-090
- 89369-008
- 89369-088
- 89369-006
- Accuracy:
- 0.01 RPM
- 0.01 RPM
- 0.01 RPM
- 0.01 RPM
- 0.01 RPM
- 0.01 RPM
- 0.01 RPM
- 0.01 RPM
- 0.01 RPM
- 0.01 RPM
- 0.01 RPM
- 0.01 RPM
- 0.01 RPM
- Drive:
- Belt driven
- Belt driven
- Belt driven
- Belt driven
- Belt driven
- Belt driven
- Belt driven
- Belt driven
- Belt driven
- Belt driven
- Belt driven
- Belt driven
- Belt driven
- Motor:
- Brushless DC
- Brushless DC
- Brushless DC
- Brushless DC
- Brushless DC
- Brushless DC
- Brushless DC
- Brushless DC
- Brushless DC
- Brushless DC
- Brushless DC
- Brushless DC
- Brushless DC
- Electrical:
- 100 - 240 V, 50/60 Hz, 35W
- 100 - 240 V, 50/60 Hz, 35W
- 100 - 240V, 50/60 Hz, 35W
- 100 - 240 V, 50/60 Hz, 35W
- 100 - 240 V, 50/60 Hz, 35W
- 100 - 240 V, 50/60 Hz, 35W
- 100 - 240 V, 50/60 Hz, 35W
- 100 - 240 V, 50/60 Hz, 35W
- 100–240V, 50/60 Hz, 35W
- 100 - 240V, 50/60 Hz, 35W
- 100 - 240V, 50/60 Hz, 35W
- 100 - 240V, 50/60 Hz, 35W
- 100 - 240V, 50/60 Hz, 35W
- Max. humidity:
- 80% up to 31 °C
- 80% up to 31 °C
- 80% up to 31 °C
- 80% up to 31 °C
- 80% up to 31 °C
- 80% up to 31 °C
- 80% up to 31 °C
- 80% up to 31 °C
- 80% up to 31 °C
- 80% up to 31 °C
- 80% up to 31 °C
- 80% up to 31 °C
- 80% up to 31 °C
- Max. capacity:
- 30 positions
- 25 positions
- 52 positions
- 15 positions
- 5 positions
- 25 positions
- 15 positions
- 5 positions
- 5 positions
- 85 positions
- 85 positions
- 52 positions
- 52 positions
- W×D×H:
- -
- -
- 790×630×1900 mm
- 790×630×650 mm
- 790×630×350 mm
- -
- 790×630×650 mm
- 790×630×350 mm
- 790×630×340 mm
- 1200×630×1900 mm
- 1200×630×1900 mm
- 790×630×1900 mm
- 790×630×1900 mm