SP Bel-Art Biohazard Disposal Bags, Bel-Art Products, a part of SP
# de catalogue 470199-972
Fournisseur: Bel-Art
Spécifications
- W×L:360×480 mm (14×19")
- Color:Red
- Description:Biohazard bags
- Height:48 cm (19")
- Thickness:1.5 mil
- Cat. no.:470199-972
- Supplier No.:F131641419
Spécifications
A propos de cet article
Bright red polypropylene bags include black biohazard symbol and precautionary procedures for added safety.
- Sterilization patch darkens when subjected to steam sterilization
- High temperature steam autoclavable to 135 °C
Certifications: Pass ASTM 1922 Tear Resistance and ASTM 1709 Dart Impact Tests.
Customers Who Bought This Also Bought