The TouchNTuff® 93-250 gloves provide enhanced grip in dry and wet environments and are the only disposable gloves that facilitate handling of oily objects. They combine the robust strength and functionality of the TouchNTuff® line with a uniquely formulated, grip-enhancing synthetic nitrile-based design.
- Touch screen capable
- Confident grip and flexibility
- Resistant to a variety of industrial chemicals
- Contains no natural rubber proteins, preventing Type I skin allergies
These single-use nitrile gloves have high tensile strength and excellent puncture resistance
Used in various industries such as automotive, chemical, food processing and services, janitorial, laboratory, life sciences (pharmaceutical, biopharmaceutical, and medical devices), manufacturing (appliances, raw materials), metal fabrication (includes glass and non-metallics), recycling and waste disposal, transportation, and warehousing.
Applications include fastening, screwing, and unscrewing parts; food product handling; handling compression press and tablet manufacturing; handling of machined parts coated with light oil; handling of paint, glue guns, and sprayers; inspection and tooling precision maintenance; lab work; blending; compounding; filling; cleaning; maintenance and equipment cleanup; picking and assembling wet or dry parts; using a V blender to mix solids and colorants; wet chemical, particle, and analytical testing.
Gloves are silicone-free, reducing the risk of product contamination from silicones.
- Catalog No:
- CA10818-396
- CA10818-404
- CA10818-402
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Disposable / reusable:
- Disposable
- Disposable
- Disposable
- Latex/Latex-free:
- Latex-free
- Latex-free
- Latex-free
- Powdered/Powder-free:
- Powder-free
- Powder-free
- Powder-free
- Finish:
- Textured fingers
- Textured fingers
- Textured fingers
- Cuff style:
- Beaded
- Beaded
- Beaded
- Finger thickness:
- 7.9 mil
- 7.9 mil
- 7.9 mil
- Palm thickness:
- 5.1 mil
- 5.1 mil
- 5.1 mil
- Anti-static:
- +
- +
- +