VWR® JerriCan
# de catalogue 75836-684
Fournisseur: VWR International
Spécifications
- Capacity:20 L
- Description:Jerrican, Rectangular, Narrow Mouth (Ø 53 mm)
- Material:HDPE
- Cat. no.:75836-684
Spécifications
A propos de cet article
These high-density polyethylene, disposable Jerri can are ideal for storing and transfer of Sterile fluids and pharmaceutical/biotech reagents.
- Excellent chemical resistance
- Leak-proof on standard test condition
- Angled-neck opening is an advantage for pouring / transfering liquid
Manufactured with 100% virgin Medical grade resin meets US FDA and USP Class VI.
Carboys have excellent chemical resistance and are well suited for use from –100 to 100 °C. Graduated to identify the volume dispensed. Comes with integrated handle for easy carrying and pouring.
Certifications: Resin meets US FDA 21 CFR, USP Class VI.
Customers Who Bought This Also Bought