Storage Box for 81 Cryo.s™ with Datamatrix, Greiner Bio-One
Supplier: Greiner Bio-One
Certificates
About this item
This storage box is designed in the class cryo box format (133 x 133 mm) for 81 Cryo.s™ with Datamatrix.
- Black rack
- Translucent high profile lid
- Sturdy polycarbonate construction
- Temperature range: -196°C to +37°C
Scanning windows underneath each tube position allow for the reading of all tube IDs in one scan. Rotation stoppers at the bottom of each tube position for usage with automated de-/recapping devices.
Customers Who Bought This Also Bought