About this item
The HyFlex® 11-751 glove features a 10G lightweight, medium-duty liner with INTERCEPT TECHNOLOGY® yarns that provide EN ISO Cut Level C protection for high levels of cut resistance.
- Silicone-free
- Increased worker confidence when handling small, sharp parts
- Exceptional abrasion resistance
The comfortable and flexible plaited liner also features an optimized Lycra® content that is soft and cool on the skin for all-day comfort
The exceptional precision, dexterity, and flexibility when handling small, sharp parts increases worker confidence while on the job. Durable and launderable with minimal shrinkage and excellent recovery after laundering, extending glove life.
INTERCEPT TECHNOLOGY® yarn has been tested to perform at or exceed industry standards. This Ansell proprietary technology offers best-in-class cut protection and increased comfort and dexterity. The palm coating design improves worker grip, extends glove life in abrasive conditions and allows the back of hand to breathe.
For use in the aerospace, automotive, machinery and equipment, and metal fabrication industries for assembly of small sharp parts, handling small sharp metal parts, light-duty stamping and maintenance.
Specifications
- Catalog No:
- 75834-210
- 75834-208
- 75834-214
- 75834-212
- 75834-206
- 75834-204
- Ambidextrous/Hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Liner material:
- Spandex/nylon
- Spandex/nylon
- Spandex/nylon
- Spandex/nylon
- Spandex/nylon
- Spandex/nylon
- Finish:
- Palm Coated
- Palm Coated
- Palm Coated
- Palm Coated
- Palm Coated
- Palm Coated
- Cuff style:
- Knit Wrist
- Knit Wrist
- Knit Wrist
- Knit Wrist
- Knit Wrist
- Knit Wrist
- EN norm:
- 4X43C
- 4X43C
- 4X43C
- 4X43C
- 4X43C
- 4X43C