Microchem® by AlphaTec™ 68-6000 Gas-Tight Chemical Protection Coveralls, Ansell
Fournisseur: Ansell Healthcare
Certificats
A propos de cet article
MICROCHEM® by AlphaTec™ 6000 is a range of lightweight, flexible and strong Level A Gas-Tight suits designed to be worn with self contained breathing apparatus to provide protection for emergency responders or chemical workers against dangerous and toxic chemicals in either liquid or gaseous form.
- Tested against numerous chemicals including the ASTM F1001 test battery
- Co-extruded multi-layer performance barrier laminate with scrim reinforcement
- Design features include a DYNAT/YKK Gas-Tight zipper that provides protection and performance in the most hostile of chemical environments
- Includes semi-rigid 3-layer PET visor providing excellent optical clarity
Disposable latex suits with ultrasonically welded and taped seams. Typically used in chemicals, pharmaceuticals, fire service, sewage treatment, nuclear, petrochemicals, first response, industrial and tank cleaning, and health service shipping.
Certifications: Tested according to EN 1073-2, EN1149-5 / AATCC 76, EN 14126 / ASTM F1671, EN943-1 Type 1a, EN943-2 Type 1a-ET.
Spécifications
- Catalog No:
- 10836-306
- Fabric:
- Chemical barrier laminate
- Seam type:
- Taped|Ultrasonically welded
- Garment application:
- Industrial, Chemical protection, Laboratory