ContactUsLinkComponent
Microflex® L22 Latex Examination Gloves, Ansell
Microflex® L22 Latex Examination Gloves, Ansell
Fournisseur:  Avantor
Certificats
Microflex® L22 Latex Examination Gloves, Ansell
Microflex® L22 Latex Examination Gloves, Ansell
Fournisseur:  Avantor
Certificats
Spécifications
  • Catalog No:
  • CA10769-354
  • Ambidextrous/Hand-specific:
  • Ambidextrous
  • Disposable / reusable:
  • Disposable
  • Latex/Latex-free:
  • Latex
  • Powdered/Powder-free:
  • Powder-free
  • Cuff style:
  • Beaded
  • Cuff thickness:
  • 3.9 mil
  • Finger thickness:
  • 5.9 mil
  • Palm thickness:
  • 5.1 mil