Microflex® L22 Latex Examination Gloves, Ansell
Fournisseur: Avantor
Certificats
A propos de cet article
Powder-free fully textured examination gloves are made using soft and supple natural rubber latex.
- Good quality rubber latex provides great barrier protection
- Exceptional elasticity makes these gloves easy to don and use over extended time
Spécifications
- Catalog No:
- CA10769-354
- Ambidextrous/Hand-specific:
- Ambidextrous
- Disposable / reusable:
- Disposable
- Latex/Latex-free:
- Latex
- Powdered/Powder-free:
- Powder-free
- Cuff style:
- Beaded
- Cuff thickness:
- 3.9 mil
- Finger thickness:
- 5.9 mil
- Palm thickness:
- 5.1 mil