3B Scientific® SONOtrain™ Ultrasound Breast Phantoms
Fournisseur: American 3B Scientific
Certificats
A propos de cet article
The material of the SONOtrain trainer simulates the feel of real soft tissue to carry out palpations and injections and shows realistic texture and echogenicity on ultrasound images.
- Easy to Use
- Cost Effective
- Realistic Echogenicity
- Available with Cysts or Tumors
- For Basic Training and Improving Dexterity
The SONOtrain trainer is an excellent teaching tool for basic training and for improving dexterity and hand-eye coordination skills.
The Phantom with cysts includes two cysts for practicing punctures and aspirations, and the Phantom with tumors contains three tumors for training on punctures or needle biopsy.
Ordering information: An ultrasound device is not part of the delivery content.
Customers Who Bought This Also Bought