3B Scientific® SONOtrain™ Gallbladder Phantom
Fournisseur: American 3B Scientific
Certificats
A propos de cet article
The material of the SONOtrain™ trainer simulates the feel of real soft tissue for carrying out palpations and injections and shows realistic texture and echogenicity on ultrasound images.
- Easy to use
- Cost effective
- Realistic echogenicity
The SONOtrain™ trainer is an excellent teaching tool for basic training and for improving dexterity and hand-eye coordination skills. The ultrasound block contains three gallbladders with different pathologies: Gallstones (diameter: 8 and 10 mm), thickened gallbladder wall and biliary sludge deposit. The gallstones and the biliary sludge deposit can move around.
Ordering information: An ultrasound device is not part of the delivery content.
Customers Who Bought This Also Bought