KIMBLE® ULTRA-WARE® Delivery Caps, DWK Life Sciences
Catalog # KT953913-0000
Supplier: DWK Life Sciences (KIMBLE)
Specifications
- Description:Delivery Cap, Three Hole, GL 45
- Cat. no.:KT953913-0000
- Supplier No.:953913-0000
Specifications
About this item
Caps have 1/4"-28 threads top and bottom with 1/8" ID holes to accept 1/8" OD PTFE tubing
Supplied complete with three 1/4"- 28 CTFE plugs and a TFE/propylene o-ring.
Customers Who Bought This Also Bought