CHROMAFLEX® Tubing with Luer Lock Adapter, Kimble Chase
# de catalogue KTH90669-0005
Fournisseur: DWK Life Sciences (KIMBLE)
Spécifications
- Tubing O.D.:¹⁄₁₆"
- Description:PTFE Tubing with Integral F-Luer CTFE Lock Adapter
- Cat. no.:KTH90669-0005
- Supplier No.:H90669-0005
Spécifications
A propos de cet article
Tubing nuts and ferrules are used to make connections between the tubing and the end fittings of CHROMAFLEX® columns.
Customers Who Bought This Also Bought