Ladies Lab Coats, Meta
Fournisseur: White Swan
Certificats
A propos de cet article
Lab coats with 65% Polyester 35% Cotton Poplin Fabric for ladies.
- Side seam slits for easy pocket access
- One Piece back and sleeve
- With three pockets
Spécifications
- Catalog No:
- 10063-684
- 10063-686
- 10063-466
- 10063-468
- 10063-676
- 10063-678
- 10063-680
- 10063-682
- 10063-464
- Garment application:
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Gender:
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- Fabric:
- 65% polyester 35% cotton Poplin fabric
- 65% polyester 35% cotton Poplin fabric
- 65% polyester 35% cotton Poplin fabric
- 65% polyester 35% cotton Poplin fabric
- 65% polyester 35% cotton Poplin fabric
- 65% polyester 35% cotton Poplin fabric
- 65% polyester 35% cotton Poplin fabric
- 65% polyester 35% cotton Poplin fabric
- 65% polyester 35% cotton Poplin fabric