CHEMFLUOR® Fluoropolymer Films, PTFE, Saint Gobain
Fournisseur: Saint Gobain Performance Plastics
Certificats
A propos de cet article
These premium grade, reusable, and easy-to-clean skived films are made from virgin PTFE
- Temperature and corrosion resistant
- Non-stick surface
- Non-porous and non-absorbent
The film will not stain or retain odors, flavors or other contaminants. Cut to size and use as weighing sheets.
The non-stick surface, temperature, and corrosion resistance features make Chemfluor film an ideal lining or covering for shelves, drawers, tables, trays, drain boards, and sinks.
Customers Who Bought This Also Bought