Graphite Ferrules for Gas Chromatography, Ohio Valley
Fournisseur: Ohio Valley
Certificats
A propos de cet article
The graphite ferrules fit “Swagelok” or “Parker” fittings
- Thermally conditioned and ready for immediate high temperature GC use
- Can be reused if not over tightened
Graphite ferrules are soft and compliant. They will
Customers Who Bought This Also Bought