Biohazard Warning Labels, Shamrock
# de catalogue 36436-200
Fournisseur: Shamrock Scientific
Small Business Enterprise
Spécifications
- Description:Biohazard warning labels
- Cat. no.:36436-200
- Dimensions:254×203.2 mm (10×8")
- Supplier No.:SBH-3
Spécifications
A propos de cet article
These Biohazard Labels comply with OSHA labeling standards for bloodborne pathogens.
Customers Who Bought This Also Bought