Adapters for Special Containers, S.C.A.T
Fournisseur: Avantor
Certificats
A propos de cet article
These adapters are designed to enable waste systems to fit JUSTRITE® containers. They can also be used to connect to CPC® coupling systems.
Customers Who Bought This Also Bought