Multi-Therm™ Benchmark Vortexer, Chemglass
Fournisseur: Chemglass
Certificats
A propos de cet article
The Multi-Therm™ Shaker is a temperature controlled vortexer that is compatible with all popular tubes and plates, including as many as six 50 mL tubes.
- Integral over temperature control ensures long life, safety and sample integrity
- Speed, time and temperature settings are continuously visible on the LCD
Used in wide applications such as denaturation of DNA, RNA and proteins, reverse transcription, lipid extractions, DNA/plasmid isolation, yeast and bacteria cultivation, gentle thawing of biological materials and restriction digests.
Spécifications
- Catalog No:
- 30623-878
- 30623-876
- Speed range:
- 200 - 1500 rpm
- 200 - 1500 rpm
- Weight:
- 8.1 kg (18 lbs.)
- 8.1 kg (18 lbs.)