Filter Degasser Receiving Flask, Microsolv
# de catalogue 89220-032
Fournisseur: MicroSolv Technology Corp
Spécifications
- Capacity:1 L (33.8 oz.)
- Description:Filter Degasser Receiving Glass Flask
- Cat. no.:89220-032
- Supplier No.:58800-47-1L