Microcentrifuge Tubes, Flat Top, Bio Plas
Fournisseur: Bio Plas
Certificats
A propos de cet article
The Bio Plas tube is built to withstand speeds over 22000 ×g
- Graduated from 0.1 to 1.5 ml
- Easy-to-open with thumb pressure
- Non wetted wall
Flat top lid forms an extra-tight seal that will not come open during processing. It features textured writing surfaces on both the lid and sides.
Customers Who Bought This Also Bought