ContactUsLinkComponent
Geissler Buret
Geissler Buret
# de catalogue 470224-052
Fournisseur:  Avantor
Geissler Buret
# de catalogue 470224-052
Fournisseur:  Avantor
Spécifications
  • Capacity:
    100 ml
Spécifications
A propos de cet article
Thick Wall Construction Borosilicate Glass.
  • Accurately graduated
  • Fitted straight bore glass body
Buret has a PTFE plug leak proof stopcock to prevent freezing, with capillary jet. Stopcock handle is plastic.
Customers Who Bought This Also Bought