Geissler Buret
# de catalogue 470224-052
Fournisseur: Avantor
Spécifications
- Capacity:100 ml
Spécifications
A propos de cet article
Thick Wall Construction Borosilicate Glass.
- Accurately graduated
- Fitted straight bore glass body
Buret has a PTFE plug leak proof stopcock to prevent freezing, with capillary jet. Stopcock handle is plastic.
Customers Who Bought This Also Bought