M121 Scepter™ Non-sterile, Lightweight, Cut Resistant Liner, Wells Lamont
Fournisseur: Wells Lamont
Certificats
A propos de cet article
These lightweight and cut-resistant liners are manufactured from polyester and stainless steel.
- Thin gauge stainless steel wrapped in white polyester
- Launderable 15 times without shrinking
- Moderate tactile sensitivity
- Latex-free
- Ambidextrous
- Packaged 10 liners per box
These non-sterile gloves can be used as a glove or liner.
Certifications: ANSI Level 4, A4
Spécifications
- Catalog No:
- 75836-520
- 75836-516
- 75836-518
- 75836-512
- 75836-514
- Model:
- M121
- M121
- M121
- M121
- M121
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Latex/Latex-free:
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Cuff style:
- Knit Wrist
- Knit Wrist
- Knit Wrist
- Knit Wrist
- Knit Wrist