Spécifications
- Ambidextrous/Hand-specific:Hand-specific
- Glove Color:White
- Glove size:L
- Length:58.4 cm (23")
- Powdered/Powder-free:Powder-free
- Latex/Latex-free:Latex-free
- Thickness:116 mil
- Cuff style:Extended
- Liner material:Nylon
- Cat. no.:32885-785
- Supplier No.:75G
Spécifications
A propos de cet article
The QRP 75G gloves are RoHS compliant gloves. PolyTuff silicon elastomer formulation offers clean, liquid-proof mid-temperature protection for wet or dry environments.
- Packaged for ISO 5 (Class 100)
- Wet or dry contact temperatures from –78 to 450 °F (–61.4 to 232 °C)
- PolyTuff silicone elastomer is sulfur-free
- Excellent resistance to chemicals, solder and fluxes
Applications of the QRP 70G Qualatherm 450 °F mid-temperature cleanroom gloves include: Diffusion furnaces, Quartz wafer boats, Burn-in ovens, PWB handling after wave soldering, ceramics, porcelains, food processing and handling.
PolyTuff silicone elastomer formulation offers clean, liquid-proof mid-temperature protection for wet or dry environments. Excellent resistance to chemicals, solders and fluxes, remaining flexible even at elevated temperatures. Seamless Comfort Curve™ paired Right Hand/Left Hand gloves minimize hand and finger fatigue. Free from PCB, asbestos and fiberglass hazards; Latex-Free. Non-particulating, cut and abrasion resistant. Micro-coated, high density, continuous filament nylon liner assures clean, easy donning and removal.