Qualatex® HiProXC 612HC Qualatex HiPro XC Latex 12" AMBI Gloves, QRP
Fournisseur: PIP
Certificats
A propos de cet article
Premium natural rubber latex gloves for ISO 5 (Class 100) applications.
- Lowest extractables and particulates
- Free from plasticizers, silicone, and powders
612HC Qualatex HiPro XC Latex Ambi Clean Room Gloves are RoHS compliant, 6 mil, 12" length, AMBI, and have textured palm and fingers.
Applications: IC and disk drive manufacture, electronics, laboratories, precision optics, assembly.
Packed 100 gloves/PBIB pack, 10 packs/case. Hi-Pro series latex gloves are manufactured free of traditional silicone oil and mold release agents which can be a primary contaminant source.
Certifications: Meets or exceeds all applicable requirements of IEST-RP-CC005.2.
Caution: This product contains natural rubber latex which may cause allergic reactions.
Spécifications
- Catalog No:
- 14003-588
- 14003-586
- 14003-592
- 14003-590
- 14003-594
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Latex/Latex-free:
- Latex
- Latex
- Latex
- Latex
- Latex
- Powdered/Powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free