Qualatherm® 55G Heat and Cold-Resistant 18" Gloves, QRP
# de catalogue 32885-936
Fournisseur: PIP
Spécifications
- Ambidextrous/Hand-specific:Ambidextrous
- Cleanroom class:100/ISO 5
- Glove Color:Brown
- Glove size:L
- Length:45.7 cm (17 7/8")
- Powdered/Powder-free:Powder-free
- Disposable / reusable:Reusable
- Liner material:Nylon
- Cat. no.:32885-936
- Supplier No.:55G
Spécifications
A propos de cet article
Designed for use in DRY heat/cold handling environments.
- Packaged for ISO 5 (Class 100)
- Dry contact temperatures for –210 to 1400 °F (–134 to 760 °C) with no charring, ash, or residue
- ESD protection Static Dissipative 10⁹
- No PCB, asbestos, or fiberglass
The 55G gloves are RoHS compliant, ambidextrous protection gloves.
Applications: Diffusion furnaces, quartz wafer boats, liquid gas flask handling (ultra cold), hot/dry glassware handling and metal fabrication.
Certifications: ESD Safe to ANSI/ESD S20.20-2014.
Customers Who Bought This Also Bought