Accessories for Reagent Reservoirs, Disposable, Sorenson Bioscience
Catalog # 89232-346
Supplier: Sorenson Bioscience
Specifications
- Color:Black
- Description:Reservoir Holder, Open Basin, SBS Footprint
- Packed:Individually wrapped
- Cat. no.:89232-346
- Supplier No.:74020
Specifications
About this item
Re-usable Reservoir Holder is on an SBS Footprint and is designed to lock reservoirs steadily and securely in place for use with the BenchTop Pipettor.
Customers Who Bought This Also Bought