ContactUsLinkComponent
VWR® High Speed Microcentrifuge
VWR® High Speed Microcentrifuge
Catalog # 75838-336
Supplier:  VWR International
VWR* Microcentrifuge 16 100xG
VWR® High Speed Microcentrifuge
Catalog # 75838-336
Supplier:  VWR International
Specifications
  • Max. capacity:
    12×1.5/2.0 ml
  • Max. speed:
    15500 rpm
  • Voltage:
    115 V
  • Description:
    High speed microcentrifuge, 115 V
  • Max. RCF:
    300 - 16100 ×g
  • Acceleration/braking time:
    11/11 s
  • Timer:
    20 s - 99 min or continuous
  • Electrical:
    115 V, 50 Hz
  • W×D×H:
    10.6×9×6.1"
  • Speed:
    300 - 15500 rpm
  • Cat. no.:
    75838-336
Specifications
About this item
The compact, affordable VWR® High Speed Microcentrifuge is capable of high speed performance up to 15500 rpm/16100×g.
  • Compact, less than 9" Ø
  • Near-silent operation
  • Large LCD with rcf conversion
  • Decelerated in just 10 seconds
  • Capacity: 12×1.5/2.0 ml
A solid, high-grade aluminum rotor is including and accepts microtubes (1.5 to 2.0 ml).
Time and speed settings are easily adjustable on a large LCD display and the speed can be set or viewed in rpm or rcf (relative centrifugal force). A safety imbalance mechanism is designed to stop the centrifuge if started out of balance and the locking lid cannot be opened unless the rotor has come to a complete stop. Other features include fast acceleration and deceleration (11 s), and reduced temperature buildup within the samples, even for extended runs at the maximum speed.
The rotor also includes a convenient snap-on plastic lid to reduce the risk of contamination.
2 year warranty.
Certifications: CE marked.
Delivery: Supplied with rotor.
Customers Who Bought This Also Bought