Market Forge Sterilmatic® STM-ED/EDX, Steam Pressure Digital Sterilizer, Crown Steam
Supplier: Market Forge
Certificates
About this item
The sterilizing cycle is fully automatic with time, temperature and venting controlled by the digital microprocessor controller. The sterilizing temperature can be set in a range from 225 °F (107 °C) and 250 °F (121°C).
- LCD display touch screen
- Temperature in °F or °C and pressure in PSI or kPa
- Sterilizing temperatures from 225 to 250°F (107 to 121 °C)
- Choose from FAST or SLOW exhaust
- Built-in USB port and RS232 serial port for optional printer
- Programable controller to store multiple temp/time/vent settings
- Maintains set temperature with built-in microprocessor
- Removable pan supports
The onboard data logger records and stores the time, temperature, and pressure for each sterilization cycle. The stored data can be either retrieved using a USB drive via the inbuilt USB port or printed out using an optional external printer. Two exhaust speeds are also available, slow and fast, depending on sterilizing media. All models are manual fill and have an electric steam trap.
Shipping weight, all models: 77 kg (169 lbs.)
Ordering information: Printer not included, this must be ordered separately.
Specifications
- Catalog No:
- 10127-644
- 10127-646
- Temperature range:
- 107...121 °C (225...250 °F)
- 107...121 °C (225...250 °F)
- Operating pressure:
- 1 atm, 100 kPa (15 psi)
- 1 atm, 100 kPa (15 psi)