ContactUsLinkComponent
GC Multiresidue Pesticide Standard #5 (34 Compounds), Restek
GC Multiresidue Pesticide Standard #5 (34 Compounds), Restek
# de catalogue 75835-054
Fournisseur:  Restek
Numéro CAS:  
GC Multiresidue Pesticide Standard #5 (34 Compounds), Restek
# de catalogue 75835-054
Fournisseur:  Restek
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Concentration:
    100 µg/ml each in toluene
  • Description:
    GC Multiresidue Pesticide Standard #5
  • Size:
    1 ml/Ampule
  • Cat. no.:
    75835-054
  • Supplier No.:
    32567
Spécifications
A propos de cet article
Accurately identify and quantify pesticide residues by GC-MS/MS in fruits, vegetables, botanicals, and herbals such as tea, ginseng, ginger, echinacea and dietary supplements.
  • Quantitatively tested to confirm composition, detailed support documentation provided
  • Organonitrogen compound
Formulated and grouped for maximum long-term stability* and well-balanced chromatographic performance.
Optimized multiresidue pesticide method is offered free of charge and downloadable XLS file includes conditions and transition tables.
Consists of 34 components, those are atrazine, bupirimate, captafol, captan, chlorfenapyr, cyprodinil, etofenprox, etridiazole, fenarimol, fipronil, fludioxonil, fluridone , flusilazole, flutriafol, folpet, hexazinone , iprodione, lenacil, mgk-264, myclobutanil, paclobutrazol, penconazole, procymidone, propargite, pyrimethanil, pyriproxyfen, tebuconazole, terbacil, terbuthylazine, triadimefon, triadimenol, tricyclazole, triflumizole and vinclozolin.
Certifications: Certified reference material (CRM) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.
Ordering information: The Comprehensive 203-compound kit also available (cat. # 32562) that covers food safety lists by the FDA, USDA, and other global governmental agencies; other ampoules also sold separately.
* NOTE: When combining many compounds with different chemical functionalities, mix stability can be an issue. In formulating these standards, we extensively studied the 203 compounds involved, and then grouped them into as few mixes as possible while still ensuring maximum long-term stability and reliability. For quantitative analysis, we recommend analysing each mix separately to ensure accurate results for every compound.
Customers Who Bought This Also Bought